Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis)
Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis)
Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum foil, 2.0mm (0.08in) thick, annealed, 99.99% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 040763.HP
also known as 040763-HP
Price (USD)
799.65
Special offer
Online exclusive
Ends: 30-Jun-2026
941.00
Save 141.35 (15%)
Each
Quantity:
100 x 500 mm
Request bulk or custom format
Price (USD)
799.65
Special offer
Online exclusive
Ends: 30-Jun-2026
941.00
Save 141.35 (15%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 2.0mm (0.08in) thick, annealed.
Assay from Supplier's CofA≥99.99% (metals basis)
Appearance (Color)Silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only