Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals
Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals
Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals
Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040769.A1
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7782-42-5
ChEBICHEBI:27594
Chemical Name or MaterialGraphite powder
ColorBlack
View more
Graphite powder is basically used for casting, polishing purposes and molding. It is also used in pigments, anodes arc-lamp carbons, coating cathode ray tubes and moderator in nuclear piles. It is used in the preparation of single sheet graphene nanoflakes and graphite based magneto rheological elastomers. It is also employed as a free standing film like cantilever sensor in piezoelectric device and additionally used as a catalyst for wet peroxide oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only