Cobalt sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Cobalt sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Cobalt sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Cobalt sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Catalog NumberQuantity
040834.KS1 Ea.
Catalog number 040834.KS
Price (USD)
368.65
Online Exclusive
409.00
Save 40.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
368.65
Online Exclusive
409.00
Save 40.35 (10%)
Each
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSputtering target, 50.8mm (2.0in) diameter x 3.18mm (0.125in) thick.
Appearance (Color)Gray
Assay from Supplier's CofA≥99.95% (metals basis)
Cobalt sputtering target is used as magnetron sputtering source, to provide a blue coloration to paints and ceramics, and also used in electroplating and its radioactive isotope, 6OCobalt, is used in the treatment of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only