Copper sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis)
Copper sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis)
Copper sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.999% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
040838.KS1 Ea.
Catalog number 040838.KS
Price (USD)
162.65
Online Exclusive
180.00
Save 17.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
162.65
Online Exclusive
180.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.999% (metals basis)
FormSputtering target, 50.8mm (2.0in) diameter x 3.18mm (0.125in) thick.
Appearance (Color)Brown
Magnetron sputtering source

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only