Chromium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
Chromium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
Chromium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Chromium sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol
Have Questions?
Catalog NumberQuantity
040845.KS1 Ea.
Catalog number 040845.KS
Price (USD)
531.65
Online Exclusive
590.00
Save 58.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
531.65
Online Exclusive
590.00
Save 58.35 (10%)
Each
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery gray
Assay from Supplier's CofA≥99.95% (metals basis)
FormSputtering target, 76.2mm (3.0in) diameter x 6.35mm (0.250in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only