Nickel sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Nickel sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Nickel sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Nickel sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.99% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
040875.KS1 Ea.
Catalog number 040875.KS
Price (USD)
288.65
Online Exclusive
321.00
Save 32.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
288.65
Online Exclusive
321.00
Save 32.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-02-0
ChEBICHEBI:28112
Chemical Name or MaterialNickel sputtering target
EINECS Number231-111-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only