Nickel Iron, sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Nickel Iron, sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

 
Have Questions?
Catalog NumberQuantity
040888.KS1 Ea.
Catalog number 040888.KS
Price (USD)
429.65
Online Exclusive
477.00
Save 47.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
429.65
Online Exclusive
477.00
Save 47.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialNickel Iron, sputtering target
Health Hazard 1H317-H351
Health Hazard 2GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only