Tin sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis)
Tin sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis)
Tin sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis)
Thermo Scientific Chemicals

Tin sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.995% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Catalog NumberQuantity
040900.KS1 Ea.
Catalog number 040900.KS
Price (USD)
370.65
Special offer
Online exclusive
Ends: 30-Jun-2026
436.00
Save 65.35 (15%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
370.65
Special offer
Online exclusive
Ends: 30-Jun-2026
436.00
Save 65.35 (15%)
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.995% (metals basis)
FormSputtering target, 76.2mm (3.0in) diameter x 3.18mm (0.125in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only