Titanium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.7% (metals basis)
Titanium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.7% (metals basis)
Titanium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.7% (metals basis)
Thermo Scientific Chemicals

Titanium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.7% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
040902.KS1 Ea.
Catalog number 040902.KS
Price (USD)
291.65
Online Exclusive
324.00
Save 32.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
291.65
Online Exclusive
324.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSputtering target, 50.8mm (2.0in) diameter x 3.18mm (0.125in) thick.
Appearance (Color)Gray
Assay from Supplier's CofA≥99.7% (metals basis)
Titanium sputtering target and its alloys are characterized by their lightness, strength and corrosion resistance and are used widely in aerospace applications. In addition, these properties also make the material suitable for medical applications (e.g. replacement hip joints). Titanium dioxide, TiO2 is used as a white pigment in paints and plastics as it provides great opacity. The same material is also used in the manufacture of heat resisting and durable glass, the TiO2 replacing certain proportions of the soda. Titanium carbide is used to manufacture cemented carbides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only