Copper gauze, 40 mesh woven from 0.056mm (0.0022in) dia wire
Copper gauze, 40 mesh woven from 0.056mm (0.0022in) dia wire
Copper gauze, 40 mesh woven from 0.056mm (0.0022in) dia wire
Thermo Scientific Chemicals

Copper gauze, 40 mesh woven from 0.056mm (0.0022in) dia wire

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 040907.FL
also known as 040907-FL
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.90
Save 10.25
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.90
Save 10.25
Each
Specifications
CAS7440-50-8
ChEBICHEBI:30052
Chemical Name or MaterialCopper gauze
EINECS Number231-159-6
IUPAC Namecopper
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only