Molybdenum gauze, 20 mesh woven from 0.305mm (0.012in) dia wire
Molybdenum gauze, 20 mesh woven from 0.305mm (0.012in) dia wire
Molybdenum gauze, 20 mesh woven from 0.305mm (0.012in) dia wire
Thermo Scientific Chemicals

Molybdenum gauze, 20 mesh woven from 0.305mm (0.012in) dia wire

Filtration, electrodes | CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
Catalog number 040908.FL
also known as 040908-FL
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
66.10
Save 9.45 (14%)
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
66.10
Save 9.45 (14%)
Each
Specifications
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum gauze
EINECS Number231-107-2
IUPAC Namemolybdenum
View more
Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only