Molybdenum gauze, 30 mesh woven from 0.204mm (0.008 in.) dia. wire, Thermo Scientific Chemicals
Molybdenum gauze, 30 mesh woven from 0.204mm (0.008 in.) dia. wire, Thermo Scientific Chemicals
Molybdenum gauze, 30 mesh woven from 0.204mm (0.008 in.) dia. wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum gauze, 30 mesh woven from 0.204mm (0.008 in.) dia. wire, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
040909.FL
also known as 040909-FL
75 x 75 mm
Catalog number 040909.FL
also known as 040909-FL
Price (USD)
73.80
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
73.80
Each
Specifications
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum gauze
EINECS Number231-107-2
IUPAC Namemolybdenum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only