Carbon disulfide, low benzene, 99.9%
Carbon disulfide, low benzene, 99.9%
Carbon disulfide, low benzene, 99.9%
Thermo Scientific Chemicals

Carbon disulfide, low benzene, 99.9%

Used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. | CAS: 75-15-0 | CS2 | 76.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
4 x 500 mL
Catalog number 040910.AY
also known as 040910-AY
Price (USD)
556.65
Special offer
Online exclusive
Ends: 30-Jun-2026
655.00
Save 98.35 (15%)
Each
Quantity:
4 x 500 mL
Request bulk or custom format
Price (USD)
556.65
Special offer
Online exclusive
Ends: 30-Jun-2026
655.00
Save 98.35 (15%)
Each
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)≥99.9%
ConcentrationBenzene: 1 ppm, max
ConcentrationHydrogen sulfide: 1.5 ppm, max
View more
Carbon disulfide is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reactive agent for the production of rubber vulcanization accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only