Molybdenum gauze, 50 mesh woven from 0.0509mm (0.002in) dia wire
Molybdenum gauze, 50 mesh woven from 0.0509mm (0.002in) dia wire
Molybdenum gauze, 50 mesh woven from 0.0509mm (0.002in) dia wire
Thermo Scientific Chemicals

Molybdenum gauze, 50 mesh woven from 0.0509mm (0.002in) dia wire

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
Catalog number 040911.FL
also known as 040911-FL
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Specifications
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum gauze
EINECS Number231-107-2
IUPAC Namemolybdenum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only