Titanium gauze, 20 mesh woven from 0.076mm (0.003in) dia wire
Titanium gauze, 20 mesh woven from 0.076mm (0.003in) dia wire
Titanium gauze, 20 mesh woven from 0.076mm (0.003in) dia wire
Thermo Scientific Chemicals

Titanium gauze, 20 mesh woven from 0.076mm (0.003in) dia wire

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
300 x 300 mm
Catalog number 040918.FL
also known as 040918-FL
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Specifications
CAS7440-32-6
ChEBICHEBI:33341
Chemical Name or MaterialTitanium gauze
EINECS Number231-142-3
IUPAC Nametitanium
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only