Titanium gauze, 50 mesh woven from 0.102mm (0.004in) dia wire
Titanium gauze, 50 mesh woven from 0.102mm (0.004in) dia wire
Titanium gauze, 50 mesh woven from 0.102mm (0.004in) dia wire
Thermo Scientific Chemicals

Titanium gauze, 50 mesh woven from 0.102mm (0.004in) dia wire

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 040921.FL
also known as 040921-FL
Price (USD)
239.65
Special offer
Online exclusive
Ends: 28-Dec-2026
282.00
Save 42.35
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
239.65
Special offer
Online exclusive
Ends: 28-Dec-2026
282.00
Save 42.35
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Carbon 0.1% max
Assay (unspecified)Oxygen 0.050% max
CommentWire diameter 0.092 - 0.112mm
Assay (unspecified)Iron 0.040% max
Appearance (Color)Silver to gray
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only