Tungsten gauze, 100 mesh woven from 0.0254mm (0.001in) dia wire
Tungsten gauze, 100 mesh woven from 0.0254mm (0.001in) dia wire
Tungsten gauze, 100 mesh woven from 0.0254mm (0.001in) dia wire
Thermo Scientific Chemicals

Tungsten gauze, 100 mesh woven from 0.0254mm (0.001in) dia wire

CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 040924.FL
also known as 040924-FL
Price (USD)
257.65
Online Exclusive
286.00
Save 28.35 (10%)
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
257.65
Online Exclusive
286.00
Save 28.35 (10%)
Each
Specifications
CAS7440-33-7
ChEBICHEBI:27998
Chemical Name or MaterialTungsten gauze
EINECS Number231-143-9
IUPAC Nametungsten
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only