Tungsten gauze, gold plated, 50 mesh woven from 0.025mm (0.001in) dia wire
Tungsten gauze, gold plated, 50 mesh woven from 0.025mm (0.001in) dia wire
Tungsten gauze, gold plated, 50 mesh woven from 0.025mm (0.001in) dia wire
Thermo Scientific Chemicals

Tungsten gauze, gold plated, 50 mesh woven from 0.025mm (0.001in) dia wire

Form: Wire Cloth; Open area: 90.3%; Width of opening: 0.019 in. | CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
Catalog number 040926.FL
also known as 040926-FL
Price (USD)
325.65
Special offer
Online exclusive
Ends: 28-Dec-2026
383.00
Save 57.35
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
325.65
Special offer
Online exclusive
Ends: 28-Dec-2026
383.00
Save 57.35
Each
Specifications
CAS7440-33-7
ChEBICHEBI:27998
Chemical Name or MaterialTungsten
EINECS Number231-143-9
IUPAC Nametungsten
View more
Filtration, electrodesTungsten gauze, gold plated, wire is used in filtration. It also serves as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only