Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis)
Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis)
Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis)
Thermo Scientific Chemicals

Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis)

Form: Wire cloth 100 x 100mm Approx.4.9g; Open area: 56.3%; Width of opening: 0.0075 in. | CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
50 x 100 mm
Catalog number 040931.FF
also known as 040931-FF
Price (USD)
389.00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
389.00
Each
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormGauze, 100 mesh woven from 0.064 (0.0025 in.) dia. wire
Total Metal Impurities≤0.01%

Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis) is used as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only