Platinum Iridium gauze, 150 mesh woven from 0.043mm (0.0017in) dia wire
Thermo Scientific Chemicals

Platinum Iridium gauze, 150 mesh woven from 0.043mm (0.0017in) dia wire

Filtration, electrodes
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 040934.FI
also known as 040934-FI
Price (USD)
-
Quantity:
50 x 50 mm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 90:10 wt%
Chemical Name or MaterialPlatinum Iridium gauze
MDL NumberMFCD02091683
Mesh Size150 Mesh
Name NoteWoven from 0.043mm (0.0017 in.) dia. wire
View more
Filtration, electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only