Stainless Steel gauze, 20 mesh woven from 0.382mm (0.015in) dia wire, Type 304
Thermo Scientific Chemicals

Stainless Steel gauze, 20 mesh woven from 0.382mm (0.015in) dia wire, Type 304

Filtration, electrodes
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 040942.HB
also known as 040942-HB
Price (USD)
167.00
Each
Quantity:
300 x 300 mm
Request bulk or custom format
Price (USD)
167.00
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni; 70:19:11 wt%
Chemical Name or MaterialStainless Steel gauze, Type 304
MDL NumberMFCD00148499
Mesh Size20 mesh
Name NoteWoven from 0.382mm (0.015 in.) dia. wire
View more
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only