Stainless Steel wire, 0.25mm (0.01in) dia, Type 304
Thermo Scientific Chemicals

Stainless Steel wire, 0.25mm (0.01in) dia, Type 304

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
150 cm
Catalog number 040945.BW
also known as 040945-BW
Price (USD)
30.20
Each
Quantity:
50 cm
Request bulk or custom format
Price (USD)
30.20
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni; 70:19:11 wt%
Chemical Name or MaterialStainless Steel wire, Type 304
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Stainless steel wire is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only