Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free
Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free
Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free
Thermo Scientific Chemicals

Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free

Solvent For U.V. Spectrophotometric and General Use | CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040976.K2
also known as 040976-K2
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.10
Save 12.45 (14%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.10
Save 12.45 (14%)
Each
Chemical Identifiers
CAS60-29-7
IUPAC Nameethoxyethane
Molecular FormulaC4H10O
InChI KeyRTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILESCCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at215nm: ≤1.00 AU
Peroxide content≤ 5ppm ( at time of packaging)
Assay (GC)≥99.0%
Optical Absorbance at230nm: ≤0.30 AU
Optical Absorbance at300nm: ≤0.005 AU
View more
Solvent for U.V. spectrophotometric and it is also important as solvent in production of cellulose plastics such as cellulose acetate. It is also common solvent for Grignard reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only