2-Propanol, GC Grade, 99.7+%
2-Propanol, GC Grade, 99.7+%
2-Propanol, GC Grade, 99.7+%
Thermo Scientific Chemicals

2-Propanol, GC Grade, 99.7+%

Suitable For Environmental, Pesticide, and GC Analysis | CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 040983.M1
also known as 040983-M1
Price (USD)
346.00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
346.00
Each
Specifications
AppearanceColorless liquid
Assay Percent Range>99.7%
Beilstein635639
Boiling Point82°C to 83°C
CAS67-63-0
View more
Suitable for environmental and GC analysis. Used in making cosmetics, skin and hair preparations, pharmaceuticals, perfumes, lacquer formulations, dye solutions, anti freezes, soaps and window cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only