Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min
Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min
Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min
Thermo Scientific Chemicals

Chloro(triphenylphosphine)gold(I), Premion™, 99.99% (metals basis), Au 39.3% min

CAS: 14243-64-2 | C18H15AuClP | 494.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
1 g
5 g
Catalog number 041007.06
also known as 041007-06
Price (USD)
937.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
937.00
Each
Chemical Identifiers
CAS14243-64-2
IUPAC Namechlorogold; triphenylphosphane
Molecular FormulaC18H15AuClP
InChI KeyIFPWCRBNZXUWGC-UHFFFAOYSA-M
SMILESCl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤0.01%
Appearance (Color)White
Assay (special)Au 39.3-39.9%
Chloro(triphenylphosphine)gold(I) is a common reagent in gold chemistry. It catalyzes certain rearrangement reactions in organic synthesis. It is also used to prepare 1-methylthyminato-N3-triphenylphosphinegold(I) by reacting with 1-methylthymine. It acts as a precursor for the synthesis of gold(I) and gold(III) organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only