Lead Tin, solder alloy, 1.6mm (0.06in) dia, with rosin core
Thermo Scientific Chemicals

Lead Tin, solder alloy, 1.6mm (0.06in) dia, with rosin core

Have Questions?
Change viewbuttonViewtableView
Quantity:
113 g
225 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041024.TB
also known as 041024-TB
Price (USD)
74.60
Each
Quantity:
113 g
Request bulk or custom format
Price (USD)
74.60
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePb:Sn; 50:50 wt%
Chemical Name or MaterialLead Tin, Solder Alloy
Health Hazard 1H302+H332-H350-H360Df-H373
Health Hazard 2GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only