Silver, solder alloy, 3.0mm (0.13in) dia
Thermo Scientific Chemicals

Silver, solder alloy, 3.0mm (0.13in) dia

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
454 g
Catalog number 041030.TD
also known as 041030-TD
Price (USD)
216.00
Each
Quantity:
454 g
Request bulk or custom format
Price (USD)
216.00
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeAg:Sn; 3.5:96.5 wt%
Chemical Name or MaterialSilver, solder Alloy
MDL NumberMFCD00801108
Name Note3.0mm (0.13 in.) dia.
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only