Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis)
Thermo Scientific Chemicals

Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis)

Evaporation source, contacts and coatings
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
5 x 10 cm
10 x 10 cm
Catalog number 041040.8G
also known as 041040-8G
Price (USD)
90.65
Online exclusive
100.00
Save 9.35
Each
Quantity:
5 x 10 cm
Request bulk or custom format
Price (USD)
90.65
Online exclusive
100.00
Save 9.35
Each
Specifications
Assay Percent NotesW ≥96.5%, Cr 99.888% (metals basis)
Chemical Name or MaterialTungsten wire, chromium plated
MDL NumberMFCD00011461
Name Note≤1.8mm (0.07 in.) dia., thick: 1.3mm (0.05 in.), Cr layer: ≤0.01 in. thick
Recommended StorageAmbient temperatures
View more
Evaporation source, contacts and coatings

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only