Tellurium(IV) ethoxide, tech. 85%, remainder ethanol
Thermo Scientific Chemicals

Tellurium(IV) ethoxide, tech. 85%, remainder ethanol

CAS: 2017-01-8 | C6H14N2O | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 041105.09
also known as 041105-09
Price (USD)
400.65
Special offer
Online exclusive
Ends: 30-Jun-2026
471.00
Save 70.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
400.65
Special offer
Online exclusive
Ends: 30-Jun-2026
471.00
Save 70.35 (15%)
Each
Specifications
Assay Percent NotesRemainder ethanol
CAS2017-01-8
Chemical Name or MaterialTellurium(IV) ethoxide
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density1.52 g/mL
View more
Tellurium(IV) ethoxide could be used in sol-gel processing for the fabrication of tellurite thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only