Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), 99.9%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), 99.9%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), 99.9%
Thermo Scientific Chemicals

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III), 99.9%

CAS: 142617-53-6 | C33H57BiO6 | 758.793 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041202.06
also known as 041202-06
Price (USD)
276.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
276.00
Each
Specifications
CAS142617-53-6
Chemical Name or MaterialTris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III)
ColorWhite to Brown
Formula Weight758.79
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only