Copper powder, -625 mesh, APS 0.50-1.5 micron, 99% (metals basis)
Copper powder, -625 mesh, APS 0.50-1.5 micron, 99% (metals basis)
Copper powder, -625 mesh, APS 0.50-1.5 micron, 99% (metals basis)
Thermo Scientific Chemicals

Copper powder, -625 mesh, APS 0.50-1.5 micron, 99% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
1 kg
Catalog number 041205.09
also known as 041205-09
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.70
Save 9.05 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 30-Jun-2026
61.70
Save 9.05 (15%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-625 mesh (less than 20 micron); APS 0.5-1.5 micron
Trace MetalsTotal metal impurities less than 1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only