alpha-Naphtholphthalein
alpha-Naphtholphthalein
alpha-Naphtholphthalein
Thermo Scientific Chemicals

alpha-Naphtholphthalein

Indicator in 0.1% or 0.04% alcohol solution | CAS: 596-01-0 | C28H18O4 | 418.448 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 041221.14
also known as 041221-14
Price (USD)
517.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
517.00
Each
Chemical Identifiers
CAS596-01-0
IUPAC Name3,3-bis(4-hydroxynaphthalen-1-yl)-1,3-dihydro-2-benzofuran-1-one
Molecular FormulaC28H18O4
InChI KeyHQHBAGKIEAOSNM-UHFFFAOYSA-N
SMILESOC1=C2C=CC=CC2=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=C2C=CC=CC2=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pink to brown
Color ChangeFrom pH 7.3 ( Pinkish - yellow) to pH 8.7 ( greenish - blue)
Specific Absorbance (E 1%/1cm)> 550 (in 0.1 N NaOH, at Lambda max)

Indicator in 0.1% or 0.04% alcohol solution.α-Naphtholphthalein is used as a pH indicator and operates in the pH ranges between 7.3 and 8.7. It is also used in the designing of optical carbon dioxide sensor based on fluorescence intensity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only