Cobalt(II) 2-methoxyethoxide, 99% (metals basis), 5% w/v in 2-methoxyethanol
Cobalt(II) 2-methoxyethoxide, 99% (metals basis), 5% w/v in 2-methoxyethanol
Cobalt(II) 2-methoxyethoxide, 99% (metals basis), 5% w/v in 2-methoxyethanol
Thermo Scientific Chemicals

Cobalt(II) 2-methoxyethoxide, 99% (metals basis), 5% w/v in 2-methoxyethanol

CAS: 142600-61-1 | C6H14CoO4 | 209.107 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 041237.AE
also known as 041237-AE
Price (USD)
323.65
Special offer
Online exclusive
Ends: 30-Jun-2026
380.00
Save 56.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
323.65
Special offer
Online exclusive
Ends: 30-Jun-2026
380.00
Save 56.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS142600-61-1
Chemical Name or MaterialCobalt(II) 2-methoxyethoxide
ColorBrown
Concentration or Composition (by Analyte or Components)5% w/v in 2-methoxyethanol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only