What is the approximate molecular weight of Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight (Cat. No. 041238.xx)?
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight, has an approximate molecular weight of 10,000-30,000 g/mol.
Search
View additional product information for Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight - FAQs (041238.14, 041238.36, 041238.22)
1 product FAQs found