Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight - FAQs
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight - FAQs
View additional product information for Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight - FAQs (041243.22, 041243.14, 041243.36)
1 product FAQs found
What is the average molecular weight of Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight (Cat. No. 041243.xx)?
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight has a typical average molecular weight of 126,000 g/mol.