Silver sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Silver sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Silver sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Silver sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.99% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
041266.KS1 Ea.
Catalog number 041266.KS
Price (USD)
1,243.00
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
1,243.00
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-22-4
ChEBICHEBI:9141
Chemical Name or MaterialSilver sputtering target
EINECS Number231-131-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only