Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis)
Thermo Scientific Chemicals

Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis)

CAS: 132951-93-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041269.03
also known as 041269-03
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.70
Save 12.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.70
Save 12.05 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS132951-93-0
Chemical Name or MaterialTin(IV) isopropoxide isopropanol adduct
Formula Weight414.15
Health Hazard 1H315-H318-H335
View more
Tin(IV) isopropoxide isopropanol adduct is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, electrochemical and conducting properties and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only