Sodium hydroxide monohydrate, 99.996% (metals basis)
Sodium hydroxide monohydrate, 99.996% (metals basis)
Sodium hydroxide monohydrate, 99.996% (metals basis)
Thermo Scientific Chemicals

Sodium hydroxide monohydrate, 99.996% (metals basis)

CAS: 12179-02-1 | H3NaO2 | 58.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 041281.06
also known as 041281-06
Price (USD)
36.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.40
Each
Chemical Identifiers
CAS12179-02-1
IUPAC Namesodium hydrate hydroxide
Molecular FormulaH3NaO2
InChI KeyJBJWASZNUJCEKT-UHFFFAOYSA-M
SMILESO.[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.
Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only