4-(2-Hydroxyethyl)benzonitrile, 98%
4-(2-Hydroxyethyl)benzonitrile, 98%
4-(2-Hydroxyethyl)benzonitrile, 98%
4-(2-Hydroxyethyl)benzonitrile, 98%
Thermo Scientific Chemicals

4-(2-Hydroxyethyl)benzonitrile, 98%

Organic intermediate | CAS: 69395-13-7 | C9H9NO | 147.177 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041283.06
also known as 041283-06
Price (USD)
873.65
Online Exclusive
970.00
Save 96.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
873.65
Online Exclusive
970.00
Save 96.35 (10%)
Each
Add to cart
Specifications
CAS69395-13-7
Chemical Name or Material4-(2-Hydroxyethyl)benzonitrile
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight147.18
Health Hazard 1H301+H311+H331-H315-H319-H335
View more
4-(2-Hydroxyethyl)benzonitrile is used to produce 4-(2-bromo-ethyl)-benzonitrile. It is used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only