Titanium slug, 9.5mm (0.37in) dia x 9.5mm (0.37in) length, 99.995% (metals basis)
Titanium slug, 9.5mm (0.37in) dia x 9.5mm (0.37in) length, 99.995% (metals basis)
Titanium slug, 9.5mm (0.37in) dia x 9.5mm (0.37in) length, 99.995% (metals basis)
Thermo Scientific Chemicals

Titanium slug, 9.5mm (0.37in) dia x 9.5mm (0.37in) length, 99.995% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 Pc.
25 Pc.
Catalog number 041303.KJ
also known as 041303-KJ
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Quantity:
5 Pc.
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormSlug, 9.5mm (0.37in) diameter.
Assay from Supplier's CofA≥99.995% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only