Chlorodiphenylphosphine, 98+%
Chlorodiphenylphosphine, 98+%
Chlorodiphenylphosphine, 98+%
Thermo Scientific Chemicals

Chlorodiphenylphosphine, 98+%

CAS: 1079-66-9 | C12H10ClP | 220.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 041352.09
also known as 041352-09
Price (USD)
32.70
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
32.70
Each
Chemical Identifiers
CAS1079-66-9
IUPAC Namechlorodiphenylphosphane
Molecular FormulaC12H10ClP
InChI KeyXGRJZXREYAXTGV-UHFFFAOYSA-N
SMILESClP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.0%

Chlorodiphenylphosphine is used in the synthesis of various phosphines and an useful reagent in for the introduction of Ph2P group. It is also useful as an intermediate to make antioxidants, flame retardants, stabilizers, catalysts, photoinitiators and optical brighteners. It is also used to produce diphenylphosphinous acid methyl esters. It is used as a precursor in the preparation of various phosphines. It is involved in the preparation of phosphines, which is used as stabilizers for plastics, flame retardants and UV-hardening paint systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only