Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)
Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)
Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)
Thermo Scientific Chemicals

Aluminum Thinfoil, 8.0 micron (0.0003in) thick, 99.0% (metals basis)

Form: approx. 0.054g/50x50mm, not certified pinhole free | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
Catalog number 041358.FI
also known as 041358-FI
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤1.0%
FormFoil, 8.0 micron (0.0003 in.) thick, not certified pinhole free
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only