1-Octadecylamine hydrochloride
1-Octadecylamine hydrochloride
1-Octadecylamine hydrochloride
Thermo Scientific Chemicals

1-Octadecylamine hydrochloride

CAS: 1838-08-0 | C18H40ClN | 305.975 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 041378.06
also known as 041378-06
Price (USD)
48.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
48.90
Each
Specifications
CAS1838-08-0
Chemical Name or Material1-Octadecylamine hydrochloride
EINECS Number217-410-2
Formula Weight305.97
Health Hazard 1H302-H315-H319-H335
View more
1-Octadecylamine hydrochloride is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only