Silver sulfate, Premion™, 99.99% (metals basis), Ag 68.9% min
Silver sulfate, Premion™, 99.99% (metals basis), Ag 68.9% min
Silver sulfate, Premion™, 99.99% (metals basis), Ag 68.9% min
Thermo Scientific Chemicals

Silver sulfate, Premion™, 99.99% (metals basis), Ag 68.9% min

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041443.14
also known as 041443-14
Price (USD)
154.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
154.00
Each
Chemical Identifiers
CAS10294-26-5
IUPAC Namedisilver(1+) sulfuric acid
Molecular FormulaAg2H2O4S
InChI KeyYPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Ag 68.9% min
Total Metal Impurities0.01% max.
Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only