Tri-n-propyltin chloride, 95%
Tri-n-propyltin chloride, 95%
Tri-n-propyltin chloride, 95%
Thermo Scientific Chemicals

Tri-n-propyltin chloride, 95%

CAS: 2279-76-7 | C9H21ClSn | 283.427 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041459.14
also known as 041459-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Specifications
CAS2279-76-7
Chemical Name or MaterialTri-n-propyltin chloride
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Density1.2678 g/mL at 28°C
EINECS Number218-910-3
View more
Tri-n-propyltin chloride is used as pharmaceutical intermediate and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only