Sodium arsenite, 0.1N Standardized Solution
Sodium arsenite, 0.1N Standardized Solution
Sodium arsenite, 0.1N Standardized Solution
Thermo Scientific Chemicals

Sodium arsenite, 0.1N Standardized Solution

CAS: 7784-46-5 | AsNaO2 | 129.909 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041533.K7
also known as 041533-K7
Price (USD)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS7784-46-5
IUPAC Namesodium oxoarsinite
Molecular FormulaAsNaO2
InChI KeyPTLRDCMBXHILCL-UHFFFAOYSA-M
SMILES[Na+].[O-][As]=O
View more
SpecificationsSpecification SheetSpecification Sheet
ConcentrationSodium arsenite 0.1N
Sodium arsenite is used as hide preservative, antiseptic, dyeing and soaps. It is used as a stressor to induce production of heat shock proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only