Potash, sulfurated, S 11.8% min as sulfide
Potash, sulfurated, S 11.8% min as sulfide
Potash, sulfurated, S 11.8% min as sulfide
Thermo Scientific Chemicals

Potash, sulfurated, S 11.8% min as sulfide

Mixture composed of potassium polysulfides and potassium thiosulfate | CAS: 39365-88-3 | K4O3S3 | 300.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 041550.22
also known as 041550-22
Price (USD)
44.50
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
44.50
Each
Chemical Identifiers
CAS39365-88-3
IUPAC Nametetrapotassium 1-oxo-1λ⁶-disulfene-1,1-bis(olate) sulfanediide
Molecular FormulaK4O3S3
InChI KeyYFKFGPGTVQHNEF-UHFFFAOYSA-L
SMILES[S--].[K+].[K+].[K+].[K+].[O-]S([O-])(=O)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥11.8% (Sulfides)
Appearance (Color)Green to yellow to brown to grey
FormGranules or chunks
Potash is used in black/brown patina (copper, brass, bronze). Its use include decorative color effects on copper, brass, bronze and nickel. Sulfurated potash has been used in the treatment of acne and other skin disorders usually in the form of a lotion with zinc sulfate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only