Titanium nitride, 99.5% (metals basis)
Titanium nitride, 99.5% (metals basis)
Titanium nitride, 99.5% (metals basis)
Thermo Scientific Chemicals

Titanium nitride, 99.5% (metals basis)

CAS: 25583-20-4 | NTi | 61.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 041556.18
also known as 041556-18
Price (USD)
88.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
88.00
Each
Chemical Identifiers
CAS25583-20-4
IUPAC Nametitanioamine
Molecular FormulaNTi
InChI KeyNRTOMJZYCJJWKI-UHFFFAOYSA-N
SMILESN#[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Titanium nitride is used as a processing agent in powder metallurgy and laboratory. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is used as Use as primary and secondary intermediate. Artificial limbs; Barrier layer in contact and interconnect metallization; Biological materials cutting tools; Gate electrode in metal-oxide-semiconductor (MOS) transistors; Low-barrier Schottky diode; Optical devices in aggressive environments; Plastic molds; Prostheses; Wear-resistant coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only