Stainless Steel foil, 0.025mm (0.001in) thick, Type 304
Thermo Scientific Chemicals

Stainless Steel foil, 0.025mm (0.001in) thick, Type 304

Fe:Cr:Ni; 70:19:11 wt%
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 041580.FI
also known as 041580-FI
Price (USD)
48.60
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
48.60
Each
Specifications
AppearanceSilver -colored foil
Chemical Name or MaterialStainless Steel
ColorSilver
Health Hazard 1H317-H350-H372
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only