Potassium phosphate, anhydrous, 97%
Potassium phosphate, anhydrous, 97%
Potassium phosphate, anhydrous, 97%
Thermo Scientific Chemicals

Potassium phosphate, anhydrous, 97%

CAS: 7778-53-2 | K3O4P | 212.265 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 041587.30
also known as 041587-30
Price (USD)
52.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.00
Save 9.35
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.00
Save 9.35
Each
Chemical Identifiers
CAS7778-53-2
IUPAC Nametripotassium phosphate
Molecular FormulaK3O4P
InChI KeyLWIHDJKSTIGBAC-UHFFFAOYSA-K
SMILES[K+].[K+].[K+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Potassium phosphate is used as a fertilizer and food additive. It serves as a buffering agent to regulate pH. It is also used in optical modulators and for non-linear optics such as second-harmonic generation (SHG). It is involved in the phase-transfer catalyzed preparation of acetylenes from the corresponding vicinal dibromo compounds through double dehydrobromination. It is also useful for the oxidative coupling of thiols to disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only